Acenaphthylene ≥94.0%
Supplier: TCI America
|
Warning
|
A0005-25G
A0005-5G
TCA0005-025GEA
145.36
CAD
TCA0005-025G
TCA0005-5G
Acenaphthylene ≥94.0%
Acenaphthylene
CAS Number: 208-96-8
MDL Number: MFCD00003806
Molecular Formula: C12H8
Molecular Weight: 152.20
Purity/Analysis Method: >94.0% (GC)
Form: Crystal
Boiling point (°C): 275
Melting point (°C): 93
Flash Point (°C): 126
Formula:
C₁₂H₈ Boiling Pt: 275 °C Melting Pt: 93 °C Density: 0.8988 g/cm³ (20 °C) Flash Pt: 126 °C |
MDL Number:
MFCD00003806 CAS Number: 208-96-8 EINECS: 205-917-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...