Phenolphthalein 0,5% in methanol 80%

Supplier: Ricca Chemical
Danger

Synonyms: 3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one

RSD54000-4A RSD54000-1A
RCSD54-1CS 597.24 CAD
RCSD54-1 RCSD54-32
Phenolphthalein 0,5% in methanol 80%
Phenolphthalein
Formula: C₂₀H₁₄O₄
MW: 318.33 g/mol
Boiling Pt: ~ 65 °C (Methanol)
Density: 0.8345 g/cm³
Flash Pt: ~ 11 °C (MeOH)
Storage Temperature: Ambient
MDL Number: MFCD00005913
CAS Number: 77-09-8
UN: 1993
ADR: 3 II
Merck Index: 14,07243
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d81b539a32ba7c40a473e6afbb8e0dae525d637&vid=91bf141a7ea82baf5df563f73967884d2a4b5d79&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR