Methyl red 1 g/L in isopropanol (IPA) indicator

Supplier: Ricca Chemical
Danger

Synonyms: 2-(4-Dimethylaminophenylazo)benzoic acid, 2-((4-(Dimethylamino)phenyl)diazenyl)benzoic acid

RSD49000-1C RSD49000-500C
RCSD49-32EA 728.38 CAD
RCSD49-32 RC-SD49.16
Methyl red 1 g/L in isopropanol (IPA) indicator
Methyl red
Formula: C₁₅H₁₅N₃O₂
MW: 269.3 g/mol
Boiling Pt: 82.5 °C [DIN 43171]
Melting Pt: –90 °C
Density: 0.8000 g/cm³
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00002425
CAS Number: 493-52-7
UN: 1219
ADR: 3 II
Merck Index: 12,06199

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR