Dichloroacetic acid (DCA) 10% in toluene, BioSyn™ detritylation reagent, for DNA/RNA synthesis, Burdick & Jackson™

Supplier: Honeywell Research Chemicals

Synonyms: DCADCA Deblock

BR676-4 BN676-20
BJBR676-4CS 2313.91 CAD
BJBR676-4 BJBN676-20
Dichloroacetic acid (DCA) 10% in toluene, BioSyn™ detritylation reagent, for DNA/RNA synthesis, Burdick & Jackson™
Dichloroacetic acid (DCA)

Detritylation Solution contains 10% Dichloroacetic acid (DCA) in Toluene (v/v), suitable for use with DNA/RNA synthesis instruments. Packaged in 20-liter steel NOWPak containers that are returned for refill. Sold only under contract.

Formula: C₂H₂Cl₂O₂
MW: 128.94 g/mol
Density: 0.9356 g/cm3
Flash Pt: 8.33 °C
MDL Number: MFCD00004223
CAS Number: 79-43-6
UN: 2924
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR