Laboratory Eye/Face Wash, Speakman®
Supplier: Speakman
SE575DP SE-575DP237 SE-575DP238
97008-318EA 5905.28 CAD
97008-318 97008-320 97008-322
Laboratory Eye/Face Wash, Speakman®
First Aid Supplies Eyewash Stations and Eyewash Bottles
Speakman® Laboratory products are designed to accommodate the needs of laboratories by offering a number of configurations to ensure the perfect fit for virtually any application.
  • Features extended arms allowing for ADA accessibility
  • Includes a universal emergency sign
  • Available in polished chrome or stainless steel finishes
These eye/face wash stations minimize the space needed for emergency equipment while offering maximum safety protection. Station 97008-318 is a wall mounted, swing-down activated eye/face wash in a recessed stainless steel cabinetMore Product Information