CELLine™ Bioreactor Flasks, Wheaton
Supplier: DWK Life Sciences (KIMBLE)
WCL1000-3 WCL1000AD-3 WCL0350-5 WCL1000-1 WCL1000AD-1 WCL0350-1
89237-440CS 2019.82 CAD
89237-440 89237-442 89237-444 89237-446 89237-448 89237-450
CELLine™ Bioreactor Flasks, Wheaton
Flasks Cell Culture Flasks
Wheaton manufactures a new line of high density cell culture membrane flasks for antibody and protein production. CELLine works well with secreting cell lines such as Hybridoma, CHO, NS0, SF, HEK, and BHK cells. Conventional in vivo or in vitro cell culture methods can be laborious, low density and require significant purification. CELLine dual compartment flasks separate the cell compartment from the bulk media compartment with a 10 kD MWCO dialysis membrane. This allows for longer run times, high density cultures, and a super concentrated harvest. The CELLine flasks were designed toMore Product Information