Vacuum Trap, [ST] Joint, Ace Glass Incorporated
Supplier: Ace Glass
8753-02 8753-04 8753-06 8753-20 8753-22 8753-24 8753-35 8753-37 8753-39 8753-40 8753-42 8753-43 8753-47 8753-49 8753-52 8753-54 8753-07 8753-09 8753-11
89063-918EA 118.58 CAD
89063-918 89063-920 89063-922 89063-936 89063-940 89063-944 89063-948 89063-950 89063-952 89063-954 89063-956 89063-958 89063-960 89063-962 89063-964 89063-966 89235-710 89235-712 89235-714
Vacuum Trap, [ST] Joint, Ace Glass Incorporated
Vacuum Traps
This vacuum trap has a [ST] outer joint on the outer body to facilitate cleaning
The inner tube has an inner [ST] joint. The trap is available with a straight tube at the top of the inner section and a straight side tube, or with a bent tube at top that is in line with the side tube, but with the side tube pointing in the opposite direction of the top tube. Fabricated from borosilicate glass.
Ordering information: Available as a complete item with outer body and inner tube, or outer body and inner tube may be purchased separately.
Order Now
SPECIFICATIONS
Environmentally Preferable