Mighty Scope Dual View Stand, Aven Inc.
Supplier: Aven
26700-215
89239-400EA 1896.02 CAD
89239-400
Mighty Scope Dual View Stand, Aven Inc.
Retort Stands
The Dual View Stand allows users to mount two Mighty Scopes
Fits Mighty Scope 1.3M and 5M.
Plug these Mighty Scopes into a PC and use the MicroViewer Software (supplied with the Mighty Scope) to view two images at the same time. Excellent for viewing a specimen from different angles. The stand offers articulating arms that allows both up and down and back and forth movements, as well as rack and pinon focusing for fine adjustments.
Working distance: 29 cm (111/2")