Thermometer, Low Temperature, Ace Glass Incorporated
Supplier: Ace Glass
8293-03 8293-07 8293-05 8293-09 8293-11 8293-04 8293-06
89062-902EA 74.48 CAD
89062-902 89062-908 89084-764 89084-770 89084-772 89062-904 89062-906
Thermometer, Low Temperature, Ace Glass Incorporated
Thermometers Analog Thermometers
Organic liquid filled themometer for use in testing procedures that normally fall below the freezing point of mercury
White back glass supplied in Total or 76mm immersion. Overall length, approximately 30cm, or 12 inches. Meets ANSI/SAMA Z236.1 standard.