Bath Oil, Ace Glass
Supplier: ACE GLASS, INC.
14115-05 14115-10 14115-12 14115-14
89078-210EA 201.93 CAD
89078-210 89078-212 89078-214 89078-216
Bath Oil, Ace Glass
Circulator Bath Fluids
An extremely stable, medium viscosity silicone heating bath oil
Will not show discoloration after 40 hours at maximum temperature.
Ordering information: Available in two temperature ranges: Low Temp — maximum 180°C; or High Temp — maximum 230°C. Low temp oil color is CLEAR. High temp oil color is AMBER.