SP Bel-Art Precisionware® Leakproof Bottles, Narrow Mouth, LDPE, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
10621-9007 10621-9015 10621-9016
47748-082CS 1183.33 CAD
47748-082 47748-084 47748-086 47748-088 47750-612 47750-614 47750-616 47750-618
SP Bel-Art Precisionware® Leakproof Bottles, Narrow Mouth, LDPE, Bel-Art Products, a part of SP
Bottles Plastic Bottles
Translucent, flexible bottles with leakproof design and matched polypropylene closures are FDA compliant and offer greater visibility than HDPE bottles.
  • Transparent, leakproof design
  • Flexible LDPE construction
  • Polypropylene closures
  • Greater visibility than HDPE bottles
  • Narrow mouth
Perfect for sampling and general storage of liquids, powders, or viscous fluids. Bottles 250 ml (8.4 oz.) and larger feature a recycling code molded into base.