Enviro-Safe® Thermometer, Organic Black Liquid Filled
Supplier: Ace Glass
8294-10 8294-11 8294-14 8294-15 8294-33 8294-34 8294-38 8294-06 8294-05 8294-04 8294-16
89062-912EA 55.93 CAD
89062-912 89062-914 89062-916 89062-918 89062-920 89062-922 89062-924 89084-784 89084-794 89062-910 89084-786
Enviro-Safe® Thermometer, Organic Black Liquid Filled
Thermometers Analog Thermometers
Enviro-Safe® lab-grade thermometers, filled with organic black liquid.
Liquid is safer since it avoids the potential hazards of mercury thermometers. Made from lead-free glass. Comes with N.I.S.T.-traceable statement of accuracy.