Stirring Shaft, Borosilicate, Ace Glass
Supplier: Ace Glass
8075-36 8075-32 8075-21 8075-14 8075-12 8075-24 8075-15 8075-23 8075-34 8075-38
89061-974EA 167.93 CAD
89061-974 89061-984 89061-972 89061-982 10099-468 89061-978 89061-976 89061-968 89061-970 89061-980
Stirring Shaft, Borosilicate, Ace Glass
Stirrers Stirring Shafts
Polished glass stirring shafts, 10 mm O.D
Can be used to join blades to button type shafts, and join agitators to beaded shafts. Plain shafts will accept agitators secured with a set screw anywhere along the ground flat section. Can also be joined to stirrer motor with the use of a chuck. Fabricated from borosilicate glass.