Upchurch Scientific® Crosses, Multi-Port Connectors, IDEX Health & Science
Supplier: Upchurch Scientific
P-723 P-635 P-634 P-722 P-730 U-430 P-729 P-777
21511-210EA 99.54 CAD
21511-210 21511-186 53500-688 21511-580 53500-816 53500-948 21511-216 53501-356
Upchurch Scientific® Crosses, Multi-Port Connectors, IDEX Health & Science
Fittings and Connectors
Upchurch Scientific® Low Pressure Tees and Crosses manufactured by IDEX Health & Science are available in two inert polymers and can handle pressures to 1,000 psi (69 bar) and each is designed with handy mounting holes
Upchurch Scientific® PEEK™ Tees and Crosses by IDEX Health & Science include high-pressure PEEK Fingertight Fittings that allows a maximum operating pressures to 3,500 psi (241 bar) when used with 1/16 in OD PEEK or stainless steel tubing.
Upchurch Scientific® MicroTees and MicroCrosses by IDEX Health and Science are used to join 1/16 in, 1/32 in, and 360 um OD capillary tubing, as well as a variety of other capillary tubing sizes using MicroTight™ tubing sleeves. All Upchurch standard MicroTees and MicroCrosses are made entirely of PEEK™ and have 0.006 in (0.150 mm) thru-holes, with resulting swept volumes ranging from 29 to 81 nL. Please Note: Use only the ferrules supplied with each connector - they are not interchangeable.
Upchurch Scientific® Stainless Steel Tees & Crosses from IDEX Health & Science are 316 stainless steel connectors that come complete with 10-32 stainless steel fittings for use with 1/16 in OD tubing and are rated to 20,000 psi (1,380 bar). They are compatible with any 10-32 coned threaded fittings.
All ETFE Tees and Crosses ship complete with 1/4-28 PFA Flangeless nuts and ETFE ferrules, while their PEEK™ polymer counterparts ship with 1/4-28 PEEK nuts and ETFE ferrules. (Products noted as "Body" versus "Assembly" do not ship with fittings).