Upchurch Scientific® Polymer Unions, IDEX Health & Science
Supplier: Upchurch Scientific
P-441 U-658 P-602 P-702 U-287 P-630 P-620 P-645 P-134 P-603 P-622 P-623 P-742 P-779-01 P-703 P-704 P-631 P-742-01
97055-086EA 44.03 CAD
97055-086 13500-024 21511-152 21511-194 21511-292 53498-560 53500-682 21511-166 21511-190 21511-104 21511-154 21511-170 21511-172 21511-224 97055-104 21511-196 21511-198 53500-684 97055-102
Upchurch Scientific® Polymer Unions, IDEX Health & Science
Fittings and Connectors
Upchurch Scientific® polymer unions are available in a variety of configurations.
  • Large Bore Unions
  • Bulkhead Unions
  • Low Pressure Unions
  • Zero Dead Volume Union Assemblies
  • NanoTight™ Unions
  • Column Couplers
Upchurch Scientific® Large Bore Unions from IDEX Health & Science allow you to use any 5/16-24 fitting and the appropriate ferrule to create a true zero dead volume connection with the P-134 Union
The U-658 Union is designed to be used with standard 1/2-20 flat-bottom threaded nuts and ferrules. Upchurch Scientific® PEEK™ Bulkhead UnionsMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable