Upchurch Scientific® Specialty Barbed Adapters, IDEX Health & Science
Supplier: Upchurch Scientific
P-757 P-767 P-799
21511-230EA 194.74 CAD
21511-230 21511-622 53501-470
Upchurch Scientific® Specialty Barbed Adapters, IDEX Health & Science
Fittings and Connectors
These unique Upchurch Scientific® tubing adapters connect peristaltic tubing to standard 1/16" or 1/8" OD tubing (using different fittings than are supplied with the product, available separately).
  • Easy to use
  • Specially designed nose is manufactured from inert PEEK™ polymer
  • Can be used as "stops" for peristaltic pump
Manufactured from inert PEEK™ polymer, a specially-designed nose allows the peristaltic tubing to simply press fit over the nose and then be held tightly in place by the retainer sleeve
These Upchurch Scientific® Conical Adapters,More Product Information