Upchurch Scientific® Barbed-to-Thread Adapters, IDEX Health & Science
Supplier: Upchurch Scientific
P-664 P-691 P-689
97055-094EA 7.93 CAD
97055-094 53501-468 97055-100
Upchurch Scientific® Barbed-to-Thread Adapters, IDEX Health & Science
Fittings and Connectors
The Upchurch Scientific® Barbed Adapters make it easy to connect flexible tubing to any standard 1/4-28 flat bottom or 10-32 coned receiving port.
  • Easy connection of tubing to flat bottom or coned receiving port
  • Three barb sizes available
  • Creates a reliable low pressure connection
Three barb sizes are available for 1/16, 1/8, and 3/16 in
Upchurch Scientific® Conical Adapters manufactured by IDEX Health & Science provide a reliable connection between 1/16 in. and 1/8 in. OD rigid/semi-rigid tubing and peristaltic-type flexible tubing, such as Tygon and PharMed for a biocompatible flow path with excellent chemical compatibility. These adapters are composed of a PEEK polymer female nut, a Super Flangeless ferrule system, and an ETFE or PEEK conical adapter body. The narrow coned end of the adapter body allows peristaltic tubing to slide on more easily than it does onto conventional barbed adapters. Peristaltic tubing is also easier to remove from Conical Adapters since no cutting or excessive pulling is required.
ID flexible tubing. Simply thread the adapter into a receiving port and slip tubing over the barbed stem to create a reliable low pressure connection.