ClipTip® Pipette Tips, 300, Thermo Scientific
Supplier: Thermo Fisher Scientific
94410510 94410513 94420513 94410517 94410518 94420518
89173-448CS 408.68 CAD
89173-448 89348-016 89348-026 89348-036 89348-046 89348-056
ClipTip® Pipette Tips, 300, Thermo Scientific
Pipette Tips
ClipTip® pipette tips provide security with a unique and innovative interlocking technology that ensures a complete seal on every channel with minimal tip attachment and ejection force.
All ClipTip® pipette tips feature a hydrophobic inner surface which effectively prevent liquid retention. ClipTip® tips are certified free of RNase, DNase, DNA, ATP, and pyrogen contamination. The tips are available in five volumes and in multiple packaging options, including environmentally friendly reload stacks.
ClipTip® 300 pipette tips suit pipetting of 30 to 300 µl volumes.
The innovative three interlocking clip design ensures the tip is held securely on the F1-ClipTip® pipette until – and only until – it is released.
Certifications: Manufactured according to ISO 9001 and ISO 13485. The F1-ClipTip® pipetting system is CE-marked in accordance with IVD Directive in Europe.