ClipTip® 50 Pipette Tips, Thermo Scientific
Supplier: Thermo Fisher Scientific
94420253 94410257 94420258 94410250 94410258
89348-022CS 1103.18 CAD
89348-022 89348-032 89348-052 89348-002 89348-042
ClipTip® 50 Pipette Tips, Thermo Scientific
Pipette Tips
ClipTip® pipette tips provide security with a unique and innovative interlocking technology that ensures a complete seal on every channel with minimal tip attachment and ejection force.
All ClipTip® pipette tips feature a hydrophobic inner surface which effectively prevent liquid retention. ClipTip® tips are certified free of RNase, DNase, DNA, ATP, and pyrogen contamination. The tips are available in five volumes and in multiple packaging options, including environmentally friendly reload stacks.
ClipTip® 50 pipette tips suit pipetting of 5 to 50 µl volumes.
The innovative three interlocking clip design ensures the tip is held securely on the F1-ClipTip® pipette until - and only until - it is released.
Certifications: Manufactured according to ISO 9001 and ISO 13485. The F1-ClipTip® pipetting system is CE-marked in accordance with IVD Directive in Europe.