ClipTip® 20 Pipette Tips, Thermo Scientific
Supplier: Thermo Fisher Scientific
94410213 94410218 94410210 94420213 94420218 94410217
89173-432CS 530.93 CAD
89173-432 89173-452 89348-000 89348-010 89348-020 89348-050 89348-030
ClipTip® 20 Pipette Tips, Thermo Scientific
Pipette Tips
Thermo Scientific ClipTip® pipette tips provide security with a unique and innovative interlocking technology that ensures a complete seal on every channel with minimal tip attachment and ejection force.
  • Pipette tips feature 'clips' that lock tips in place and forms a complete seal
  • Every ClipTip pipette tip utilises low retention technology to enable maximium sample recovery
  • Certified free of RNase, DNase, DNA, ATP and endotoxin contamination
  • Each tip insert has ink-jetted lot and volume information; the convenient colour key makes it easy to match the
More Product Information