Upchurch Scientific® Thread-to-Thread Adapters, IDEX Health & Science
Supplier: Upchurch Scientific
1583 U-510 P-671 P-627 P-770 1591 U-659 P-621 P-650 1592 P-652 U-500-01 U-504 P-135 P-881 P-882 P-626 U-511 P-669-01 UH-630
53498-526EA 76.03 CAD
53498-526 97055-124 13500-478 21511-180 53403-086 53500-702 13500-028 21511-168 21511-192 21511-416 53500-766 21511-690 21528-168 53498-550 97055-108 97055-110 21511-178 21511-692 97055-096 97055-134
Upchurch Scientific® Thread-to-Thread Adapters, IDEX Health & Science
Fittings and Connectors
IDEX Health & Science offers a wide assortment of Upchurch Scientific® threaded port adapters to help bring products with different thread configurations together.
  • High pressure ratings
  • Variety of adapters available
  • Choose from polymer and stainless steel versions
The pressure ratings of these adapters often exceed the pressure holding ability of the fittings and tubing used with them
Create a true zero dead volume connection between 1/16 in. capillary tubing with Upchurch Scientific® MicroTight™ Adapters from IDEX Health & Science in an inline connection with the ability to withstand up to 5,000 psi (345 bar), depending upon the adapter configuration. (When MicroTight tubing sleeves are used, the pressure rating lowers to 4,000 psi / 276 bar).
External National Pipe Thread Adapters make connections to female 1/8 in and 1/4 in National Pipe Thread (NPT) ports. Manufactured from PEEK™ polymer by IDEX Health & Science, Upchurch Scientific® NPT Adapters are durable and chemically resistant and available in a variety of female threads, plus a male threaded version that is suitable for most low pressure applications. Please Note: Wrap the threads on the NPT side of these adapters with thread seal tape (plumbers tape) to ensure a leak-free seal.
Choose from polymer and stainless steel versions.