Glove Dispenser With 4 Compartments And Front Access, S-Curve Technologies
Supplier: S-Curve Technologies
GD-400 GD-400 PETG
32948-900EA 1201.65 CAD
32948-900 98106-918
Glove Dispenser With 4 Compartments And Front Access, S-Curve Technologies
Glove Dispensers and Glove Box Holders
Glove dispenser with 4 compartments, individual lids and front access.
Wall mounts or free-standing-wall mount bracket and support bar included.
Flame polished edges for smoothness.
Order Now
SPECIFICATIONS
Environmentally Preferable