Fiberlite™ H3-LV Large Volume Swinging Bucket Rotor, Thermo Scientific
Supplier: Thermo Scientific
75003665 75003665
CA97040-280EA 52985.93 CAD
CA97040-280 97040-280
Fiberlite™ H3-LV Large Volume Swinging Bucket Rotor, Thermo Scientific
Centrifuges Rotors
Thermo Scientific* Fiberlite H3-LV Rotor is for high-volume microplate achieves high-volume microplate processing.
These lightweight, carbon fiber rotors maximize performance with versatility, speed and a robust, corrosion-free design.
Enhanced productivity: Reduce run-times with higher G-forces with our lightweight carbon fiber rotors.
Unequalled durability: Corrosion-and fatigue-resistant design ensures structural integrity.
Secure investment: Backed by a 15-year warranty.
Recommended for Pelleting cells and cellular debris, protein precipitation, plasmid purification and collecting physiological fluids for diagnostic tests.
Ordering information: Compatible with Thermo Scientific Nalgene bottles and tubes for complete centrifuge solutions. Compatible for Thermo Scientific Multifuge X3/X3R/X3F/X3FR Centrifuge Series.