Fiberlite™ F15-8x50cy Fixed-Angle Rotor, Thermo Scientific
Supplier: Thermo Scientific
75003663 75003663
CA97040-276EA 34268.68 CAD
CA97040-276 97040-276
Fiberlite™ F15-8x50cy Fixed-Angle Rotor, Thermo Scientific
Centrifuges Rotors
Thermo Scientific's Fiberlite F15-8x50cy Fixed-Angle Rotor pellets fractional samples with high RCF
  • Reduced run-times with higher G-forces achieved with lightweight carbon fiber rotors
  • Corrosion- and fatigue-resistant design that ensures structural integrity
  • Backed by a 15-year warranty
Recommended for pelleting cells, bacteria, subcellular fractions, and small amounts of nucleic acids and viruses.
The lightweight, carbon fiber rotor maximizes performance with versatility, speed and a robust, corrosion-free design.
Certifications: Biocontainment certified by CAMR, HPA, Porton Down, U.K.
Ordering information: Adapters are sold separately. O-ring Refresher Kit (021-085-077) is also available. Contact your Sales Representative for more information. Compatible with Thermo Scientific Multifuge X1/X3/X3 F Series.