Bel-Art Magic Touch 2™ Ice Pans, ATS Life Sciences
Supplier: Bel-Art Products, a Part of SP
M16807-1101 M16807-1102 M16807-1103 M16807-1104 M16807-4101 M16807-4102 M16807-4103 M16807-4104 M16807-9101 M16807-9102 M16807-9103 M16807-9104
89233-332EA 161.39 CAD
89233-332 89233-334 89233-336 89233-338 89233-356 89233-358 89233-360 89233-362 89233-364 89233-366 89233-368 89233-370 89233-372 89233-374 89233-376 89233-378
Bel-Art Magic Touch 2™ Ice Pans, ATS Life Sciences
Ice Containers
Superior insulating properties combined with high chemical resistance make Magic Touch 2™ Ice Pans compatible not only with wet ice, but also with ultra-low temperature materials including dry ice, salt slurries, dry ice solvent slurries, or liquid nitrogen.
  • Excellent insulation
  • High chemical resistance
  • Convenient pour spout
  • Stackable
  • Lightweight and durable
  • Expanded urethane construction
  • Temperature range: –196...+100 °C (–320... +212 °F)
Able to withstand a wide temperature range, these high-quality, expanded urethane, laboratory ice pans are lightweight, durable, non-sweating and impervious to moisture and odors.
Ordering information: Magic Touch 2™ Ice Pans are available in 1.0 L Mini, 4.0 L Midi, and 9.0 L Maxi sizes, each in four colors; black, red, blue, and green.