VWR® Heated Circulating Baths, 240V Models
Supplier: VWR International
89202-928 89202-944 89400-972 89400-976
89202-928EA 5731.8 CAD
89202-928 89202-944 89400-972 89400-976
VWR® Heated Circulating Baths, 240V Models
Circulating Thermostats Open Circulator Systems Baths with Integral Circulators
VWR® Heated Circulating Baths offer precise temperature control, at temperature ranges from ambient +10° to 200°C (depending on the controller).
  • Easy-to-read LCD display
  • Programmable for up to ten 100-step programs
  • Available in a wide range of sizes
Available in a wide range of sizes, with three controller options, these heated baths are ideal for many applications.
The large LCD display on the MX temperature controller is easily viewed across the lab. The Advanced Digital temperature controller offers a touch-pad, while the Advanced Programmable Temperature Controller includes a full touch-screen display, and programmability (ten 100-step programs).
The Advanced Series Controllers offer Swivel 180™ technology, which allows you to rotate the controller independently from the bath. All heated circulating baths include LidDock™ lid stowing system and DuraTop™, the precision molded phenolic bath top, which remains cooler at high temperatures and resists laboratory chemicals.
Certifications: ETL listed. CE marked. MX Controller complies with DIN 12876-1 Class I safety requirements for use with non-flammable liquids. Advanced Digital and Advanced Programmable comply with DIN 12876-1 Class III safety requirements for use with flammable liquids. cETLus compliant.
Ordering information: Heated Circulators include: reservoir cover; bypass tubing; male inlet and outlet adapters for 3/8", 1/4" and 3/16" (.95, .63 and .47 cm) tubing. 1/4" NPT to M16 adapters also included on 50 Hz models.
Caution: Note: These units are all 240 V, 50 Hz units. Corresponding 120 V models are also available, these are: 89202-926, 89202-942, 89400-970 and 89400-974 respectively.