NxGen™ phi29 DNA Polymerase, Biosearch Technologies
Supplier: LUCIGEN CORPORATION CA
30221-1 30221-2
CA97065-180EA 1630.02 CAD
CA97065-180 CA97065-182
NxGen™ phi29 DNA Polymerase, Biosearch Technologies
Nucleic Acid Reagents End-point PCR Enzymes and Kits
Lucigen’s NxGen™ phi29 DNA Polymerase is designed for excellent strand displacement, fidelity, and speed
phi29 DNA Polymerase is a highly processive DNA polymerase with a powerful strand displacement activity and a 3′→ 5′ proofreading exonuclease function. The enzyme is capable of up to 70,000 insertions per binding event.
Performance Specifications: 1) Unit Concentration: 10,000 U/mL. 2) Purity (SDS-PAGE): >99%. 3) SS Exonuclease: Functional. 4) E. coli 16S rDNA Contamination: 100 U <10 copies. 5) Endonuclease: 100 U <10% converted. 6) Storage: -20°C.
Source: A recombinant E. coli strain carrying the phi29 DNA Polymerase gene from bacteriophage phi29.
Ordering information: Supplied at 10,000 U/mL in 10 mM Tris-HCl, 100 mM KCl, 0.1 mM EDTA, 1 mM dithiothreitol, 0.5% Tween-20, 0.5% NP-40, 50% glycerol, pH 7.4 @ 25°C. Also supplied is 10X phi29 DNA Polymerase Buffer composed of 500 mM Tris-HCl, 100 mM (NH4)2SO4, 40 mM Dithiothreitol, 100 mM MgCl2, pH 7.5 @ 25°C. 10,000 U is supplied as 5 × 2,000 U. Unit definition: 1 unit is defined as the amount of polymerase required to convert 0.5 pmol of dNTP's into acid insoluble material in 10 minutes at 30°C. Note: This product is sold for in vitro research use only under license from Brookhaven National Laboratory, U.S. Patent Nos. 4,952,496; 5,693,489; and 5,869,320.