Generator Probe Tool Kits, Omni International
Supplier: Omni International
10505 10507 T1001 10506 17104 10504 10503
CA11010-706PK 733.39 CAD
CA11010-706 CA11010-708 CA11010-712 CA11010-704 CA11011-604 CA11010-710 CA11010-622
Generator Probe Tool Kits, Omni International
Tool Sets
Stainless Steel Generator Probes are configured with a PTFE upper and lower bearing.
Specially designed Generator Probe Tool Kits are required to disassemble probes for cleaning and bearing replacement. Use of any other tools improper bearing maintenance could damage the generator probe.
Over time, PTFE bearings can wear out and must be replaced. The most common signs of worn bearings are: If the bearing no longer fits snugly against the rotor shaft, when visible wear is apparent or if black particles become visible in the sample.
Delivery information: Each bearing package contains 4 bearings.