Automation Pipette Tips for Tecan Te-MO and Freedom EVO Workstations, Thermo Scientific
Supplier: Thermo Scientific
162-96R 162-96RS 163-96R 163-96RS LMF163-96RS-50 161-96RS 906-021 907-261 907-262
14004-618CS 1305.48 CAD
14004-618 14004-620 14004-624 14004-626 14004-628 14004-613 72830-586 72830-588 72830-590
Automation Pipette Tips for Tecan Te-MO and Freedom EVO Workstations, Thermo Scientific
Pipette Tips
Thermo Scientific provides a solution for your Tecan robotic pipette tips.
These automation pipette tips have been designed, validated, and tested on a Tecan EVO robot to ensure equal performance without the need to modify your existing protocols and programs. Our low percentage Coefficient of Variation (%CV) ensures product consistency and performance from every lot. Each lot must pass a 15-point QC inspection including performance testing on our Tecan workstation before tips are released. All tips are certified RNase, DNase, DNA, endotoxin, bioburden, and pyrogen free.