Drum Spill Stations, SECURALL®
Supplier: Securall Cabinets
DSS02 DSS04 DSS06 DSS09 DSS12
89237-804EA 3575.68 CAD
89237-804 89237-806 89237-808 89237-810 89237-812
Drum Spill Stations, SECURALL®
Drum Storage
Controlling unwanted spills is easier than ever with SECURALL® Drum Spill Pallets
Built to provide a lifetime of service, these secondary containment Drum Spill Stations provide an easy and safe solution to organizing hazardous waste in drums. Rugged all-welded 12-gauge galvannealed steel offers long term use. Our Drum Spill Stations are specifically designed to be used as a holding area for drums, dispensing station or waste collection center. All units feature steel planking and forklift channels for easy mobility. Durable enough to be transported at full capacity.
Certifications: SMaRT Certified; Complies with EPA Spill Containment Requirements (40CFR264.175).
Ordering information: Standard Color is Gray, please contact your Avantor Furniture Specialist for more information.