Hazmat Storage Buildings, SECURALL®
Supplier: Securall Cabinets
B200 B400 B600 B900 B1200 B1600 B2400 B3200 B3800 B4800 B6400 B8000
89237-638EA 18569.03 CAD
89237-638 89237-640 89237-642 89237-644 89237-646 89237-648 89237-650 89237-652 89237-654 89237-656 89237-658 89237-660
Hazmat Storage Buildings, SECURALL®
Furniture Storage Cabinets
SECURALL® Hazmat Storage Buildings are designed for Flammable Liquid Storage, Chemical Storage, and Hazardous Materials in 55-gallon drums.
  • Additional options are available
Please contact your Avantor Furniture Specialist for more information. NOTE: Building may require State Approval. Please check with your Local Authorities (i.e.: Fire Marshall) to determine if State Approval and/or State Seal are required. Additional charges and lead times may apply.
Securall® drum storage buildings / lockers are available in many standard models to help our customers gain OSHA and EPA compliance of your hazardous chemical storage.
Certifications: SMaRT Certified; FM Approved.
Ordering information: Standard Color is Gray, please contact your Avantor Furniture Specialist for more information.