Tungsten ICP Standard, 1 ml = 10 mg W (10000 ppm W)W in 5% HNO3 / 2% HF, RICCA Chemical Company
Supplier: Ricca Chemical
RPW10KN0-100N RPW10KN0-50N
RCPW10KN-100EA 374.46 CAD
RCPW10KN-100 RCPW10KN-50
Tungsten ICP Standard, 1 ml = 10 mg W (10000 ppm W)W in 5% HNO3 / 2% HF, RICCA Chemical Company
Standards Cationic Standards Tungsten Standards