Palladium(II) nitrate (0.5% Pd) in nitric acid 5% matrix modifier solution
Supplier: Ricca Chemical
|
Danger
|
RMMPD5KN-500N
RMMPD5KN-50N
RMMPD5KN-100N
RCMMPD5KN-50EA
795.53
CAD
RCMMPD5KN-500
RCMMPD5KN-100
RCMMPD5KN-50
Palladium(II) nitrate (0.5% Pd) in nitric acid 5% matrix modifier solution
Palladium (II) nitrate
Formula:
N₂O₆Pd MW: 230.43 g/mol Boiling Pt: ~ 100 °C Melting Pt: ~ 0 °C Density: 1.0500 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00011169 CAS Number: 10102-05-3 UN: 3264 ADR: 8,III |
Specification Test Results
Appearance | Clear, brown, odorless |
Certified Concentration | 5,000 ± 50 ppm Pd |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...