Palladium(II) nitrate (1.0% Pd) in nitric acid 5% matrix modifier solution

Supplier: Ricca Chemical
Danger

RMPD10KN-50N
RCMMPD10KN-50EA 1702.7 CAD
RCMMPD10KN-50
Palladium(II) nitrate (1.0% Pd) in nitric acid 5% matrix modifier solution
Palladium (II) nitrate
Formula: N₂O₆Pd
MW: 230.43 g/mol
Boiling Pt: ~ 100 °C
Density: 1.0400 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011169
CAS Number: 10102-05-3
UN: 3264
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear, brown, odorless
Certified Concentration 10,000 ± 500 ppm Pd

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR