Zirconyl-SPADNS reagent for fluoride determination

Supplier: Ricca Chemical
Danger

R9850000-500C R9850000-1C
RC985016EA 126.4 CAD
RC985016 RC985032
Zirconyl-SPADNS reagent for fluoride determination
Zirconyl-SPADNS reagent
Boiling Pt: ~ 102 °C
Density: 1.1000 g/cm³
Storage Temperature: Ambient
UN: 1789
ADR: 8,II

Order Now

Specification Test Results

Appearance Very dark, reddish tint, slightly pungent odor

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR