Zinc sulfate 1.180 Specific Gravity for concentration of intestinal parasites, eggs, and cysts

Supplier: Ricca Chemical

R9650000-4A R9650000-500A R9650000-1A
RC96501EA 371.3 CAD
RC96501 RC965016 RC965032
Zinc sulfate 1.180 Specific Gravity for concentration of intestinal parasites, eggs, and cysts
Zinc sulphate
Formula: O₄SZn
MDL Number: MFCD00011302
CAS Number: 7733-02-0
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Specific Gravity 1.175 ± 0.005 sp gr at 25.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR