Zinc acetate 2 N

Supplier: Ricca Chemical

R9450000-500A R9450000-120A
RC945016EA 148.52 CAD
RC945016 RC94504
Zinc acetate 2 N
Zinc acetate

220 g + 870 ml H₂O

Formula: C₄H₆O₄Zn
MW: 183.48 g/mol
MDL Number: MFCD00012454
CAS Number: 557-34-6
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Refractive Index Confirmed concentration

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR