Wright-Giemsa stain

Supplier: Ricca Chemical
Danger

R9380000-500C R9380000-1C
RC938016EA 153.26 CAD
RC938016 RC938032
Wright-Giemsa stain
Wright-Giemsa stain (solutions & kits)

Wright-Giemsa Stain Mixture

Boiling Pt: ~ 64.5 °C
Density: 0.8484 g/cm³
Flash Pt: ~ 12 °C (MeOH)
Storage Temperature: Ambient
MDL Number: MFCD00132593
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Absorbance (traceable to NIST SRM 930) Conforms to control
Appearance Dark blue, Methanol odor


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR