Wright's stain for staining and general differentiation of blood corpuscles, regular formula

Supplier: Ricca Chemical
Danger

R9360000-4C R9360000-500C R9360000-1C
RC93601EA 567.22 CAD
RC93601 RC936016 RC936032
Wright's stain for staining and general differentiation of blood corpuscles, regular formula
Wright's stain
Boiling Pt: ~ 64.5 °C
Density: 0.8000 g/cm³
Flash Pt: ~ 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00082143
CAS Number: 68988-92-1
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Absorbance (traceable to NIST SRM 930) Conforms to control
Appearance Dark blue, Methanol odor
pH at 25 °C (traceable to NIST SRM 185 & 186) 6.4 ± 0.1 pH at 25.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR