Wright's stain for staining and general differentiation of blood corpuscles, rapid formula
Supplier: Ricca Chemical
|
Danger
|
R9350000-4C
R9350000-500C
R9350000-1C
RC93501EA
639.9
CAD
RC93501
RC935016
RC935032
Wright's stain for staining and general differentiation of blood corpuscles, rapid formula
Wright's stain
Boiling Pt:
~ 64.5 °C Density: 0.8000 g/cm³ Flash Pt: ~ 12 °C Storage Temperature: Ambient |
MDL Number:
MFCD00082143 CAS Number: 68988-92-1 UN: 1993 ADR: 3,II |
Specification Test Results
Absorbance (traceable to NIST SRM 930) | Conforms to control |
Appearance | Dark blue, Methanol odor |
pH at 25 °C (traceable to NIST SRM 185 & 186) | 6.4 ± 0.1 pH at 25.0 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...