Water Hardness Buffer, with Magnesium EDTA, Ricca Chemical Company
Supplier: Ricca Chemical
R9200000-4A
R9200000-20F
RC92001EA
388.68
CAD
RC92001
RC9200-5
Water Hardness Buffer, with Magnesium EDTA, Ricca Chemical Company
Buffers
Biological Buffers
Suitable for use in the following compendial methods:
-ASTM D511 A, Reagent 11.1
-APHA 2340 C, Reagent 2a1
-EPA 215.2, Reagent 5.3.6
-EPA 130.2, Reagent 6.1.1