Bathocuproine 0.1% (w/v) in aqueous solution

Supplier: Ricca Chemical

R0905000-500A R0905000-120A
RC90516EA 1463.08 CAD
RC90516 RC9054
Bathocuproine 0.1% (w/v) in aqueous solution
Bathocuproine
Boiling Pt: 100 °C
Melting Pt: 0.0 °C
Density: 1.0000 g/cm³
Storage Temperature: Ambient
CAS Number: 52698-84-7

Order Now

Specification Test Results

Appearance Clear, colorless, no odor


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR