Vanadomolybdate reagent for phosphorus determination

Supplier: Ricca Chemical

R8980000-500A R8980000-1A
RC898016EA 126.4 CAD
RC898016 RC898032
Vanadomolybdate reagent for phosphorus determination
Vanadomolybdate reagent
UN: 2031
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear, yellowish-green, Hydrochloric odor
Suitability for Use Yellow in the presence of Phosphate


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR