Turk's solution, diluting fluid for leukocyte count
Supplier: Ricca Chemical
Danger
R8850000-500A R8850000-1A
RC885016EA 71.1 CAD
RC885016 RC885032
Turk's solution, diluting fluid for leukocyte count
Turk's solution
For In Vitro Diagnostic Use. Suitable for use as a diluting fluid for Leukocyte (white blood cell) counting.
IVDLogo
Order Now
Specification Test Results
Absorbance (traceable to NIST SRM 930) Conforms to control
Appearance Purple, odorless
SPECIFICATIONS
Environmentally Preferable