Thioacetamide 0,25% (w/v) in aqueous solution

Supplier: Ricca Chemical
Danger

Synonyms: Ethanethioamide

R8393000-500A R8393000-120A
RC839316EA 135.88 CAD
RC839316 RC83934
Thioacetamide 0,25% (w/v) in aqueous solution
Thioacetamide
Formula: CH₃CSNH₂
MW: 75.13 g/mol
Boiling Pt: 100 °C
Melting Pt: 0.0 °C
Density: 1.0010 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD00008070
CAS Number: 62-55-5
Merck Index: 13,09391
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe2000e62db991c85340c257ac6d64ca42c3551a&vid=18e76b2cdee9ee22a2618ec371858a498f218457&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear, colorless, odorless to slight mercaptan odor


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR