L(+)-Tartaric acid 2% (w/v) in aqueous solution

Supplier: Ricca Chemical
Warning

Synonyms: L-Tartaric Acid, (2R,3R)-(+)-Tartaric acid, Tartaric Acid, (R,R)-(+)-2,3-Dihydroxysuccinic acid, (R,R)-(+)-Tartaric acid, (2R,3R)-2,3-Dihydroxysuccinic acid

R8370000-500A R8370000-1A
RC8370-16EA 436.08 CAD
RC8370-16 RC8370-32
L(+)-Tartaric acid 2% (w/v) in aqueous solution
L(+)-Tartaric acid
Formula: C₄H₆O₆
MW: 150.09 g/mol
Boiling Pt: 399
Melting Pt: 0.0 °C
Density: 1.0100 g/cm³
Flash Pt: 150
Storage Temperature: Ambient
MDL Number: MFCD00064207
CAS Number: 87-69-4
Merck Index: 13,09158
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d4e859e52081482a351d52322717614aad450742&vid=879c292903ddee7a59d5d6b2d969c4c8fc6eac75&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Refractive Index Confirmed Concentration


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR