Silver sulfate solution in sulphuric acid for COD determination

Supplier: Ricca Chemical
Danger

R8350000-500C1 R8350000-1C1
RC835016EA 431.34 CAD
RC835016 RC835032
Silver sulfate solution in sulphuric acid for COD determination
Silver sulphate
Formula: Ag₂SO₄
Boiling Pt: ~ 338 °C
Density: 1.8000 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003407
CAS Number: 10294-26-5
UN: 1830
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear, colorless, odorless


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR