Sulfuric acid 0.6667 N (2N/3) suitable for preparation of protein-free filtrates of blood, serum, or plasma

Supplier: Ricca Chemical

R8290000-1A R8290000-20F R8290000-4A
RC8290-32EA 293.88 CAD
RC8290-32 RC8290-5 RC8290-1
Sulfuric acid 0.6667 N (2N/3) suitable for preparation of protein-free filtrates of blood, serum, or plasma
Sulfuric acid
Formula: H₂SO₄
MW: 98.08
Boiling Pt: 330
Melting Pt: 10.38
Density: 1.84
MDL Number: MFCD00064589
CAS Number: 7664-93-9
UN: 1830
ADR: 8 II

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 723) 0.6667 ± 0.0005 N at 20.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR