Sulfuric acid ≥80% (v/v) in aqueous solution

Supplier: Ricca Chemical

R8190000-500C1 R8190000-1C1
RC819016EA 794.74 CAD
RC819016 RC819032
Sulfuric acid ≥80% (v/v) in aqueous solution
Sulfuric acid
Formula: H₂SO₄
MDL Number: MFCD00064589
CAS Number: 7664-93-9
UN: 1830
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 84) 80.0 ± 0.5 % (v/v)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR