Sulfuric acid 16.7% (v/v) in aqueous solution
Supplier: Ricca Chemical
R8162000-500A
R8162000-1A
RC8162-16EA
158
CAD
RC8162-16
RC8162-32
Sulfuric acid 16.7% (v/v) in aqueous solution
Sulfuric acid
Formula:
H₂SO₄ |
MDL Number:
MFCD00064589 CAS Number: 7664-93-9 UN: 1830 ADR: 8,II |
Specification Test Results
Appearance | Clear, colorless, odorless |
Assay at 20 °C (traceable to NIST SRM 84) | 16.67 ± 0.03 % (v/v) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...